Crotonoyl chloride cis- and trans-mixture ≥95.0%
Supplier: TCI America
Synonyms:
But-2-enoyl chloride, 2-Butenoyl chloride, Crotonoyl Chloride, Crotonic acid chloride
C0424-25G
C0424-500G
TCC0424-025GEA
56
USD
TCC0424-025G
TCC0424-500G
Crotonoyl chloride cis- and trans-mixture ≥95.0%
Crotonoyl chloride
CAS Number: 10487-71-5
MDL Number: MFCD00000734
Molecular Formula: C4H5ClO
Molecular Weight: 104.53
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 35
Specific Gravity (20/20): 1.08
Formula:
C₄H₅ClO MW: 104.53 g/mol Boiling Pt: 125 °C Density: 1.08 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00000734 CAS Number: 10487-71-5 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...