Croton oil

Supplier: TCI America

Synonyms: Tiglium oil

C0421-25ML C0421-5ML
TCC0421-025MLEA 191.4 USD
TCC0421-025ML TCC0421-5ML
Croton oil
Croton oil

CAS Number: 8001-28-3
MDL Number: MFCD00130880
Form: Clear Liquid
Color: Yellow Red
Flash Point (°C): 74
Specific Gravity (20/20): 0.94


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Lipids

Density: 0.94 (20 °C)
Flash Pt: 74 °C
MDL Number: MFCD00130880
CAS Number: 8001-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR