Disodium hydrogen citrate sesquihydrate ≥98.0% for synthesis
Supplier: TCI America
Synonyms:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
C0365-25G
C0365-500G
TCC0365-025GEA
20
USD
TCC0365-025G
TCC0365-500G
Disodium hydrogen citrate sesquihydrate ≥98.0% for synthesis
di-Sodium hydrogen citrate sesquihydrate
CAS Number: 6132-05-4
MDL Number: MFCD00150445
Molecular Formula: C6H8O7
Molecular Weight: 236.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 149
Other Product Names / Synonyms: Citric Acid Disodium Salt, Disodium Hydrogen Citrate
Formula:
C₆H₆Na₂O₇·1.5H₂O Melting Pt: 149 °C |
MDL Number:
MFCD00150445 CAS Number: 6132-05-4 EINECS: 242-734-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...