4-Chloro-o-toluidine ≥98.0% for synthesis
Supplier: TCI America
|
Danger
|
Synonyms:
2-Amino-5-chlorotoluene, 4-Chloro-2-methylaniline, 4-Chloro-o-toluidine, Fast Red TR Base
A0704-25G
A0704-500G
TCA0704-025GEA
29
USD
TCA0704-025G
TCA0704-500G
4-Chloro-o-toluidine ≥98.0% for synthesis
4-Chloro-o-toluidine
CAS Number: 95-69-2
MDL Number: MFCD00007842
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 241
Flash Point (°C): 124
Freezing point (°C): 25
Other Product Names / Synonyms: 2-Amino-5-chlorotoluene, 4-Chloro-o-toluidine, Fast Red TR Base
Formula:
C₇H₈ClN Boiling Pt: 241 °C Melting Pt: 27.2 °C Density: 1.14 g/cm³ (20 °C) Flash Pt: 124 °C |
MDL Number:
MFCD00007842 CAS Number: 95-69-2 EINECS: 202-441-6 UN: 2239 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...