2'-Ethoxyacetanilide ≥98.0%
Supplier: TCI America
Synonyms:
N-Acetyl-o-phenetidine
A0103-25G
A0103-500G
TCA0103-025GEA
61.7
USD
TCA0103-025G
TCA0103-500G
2'-Ethoxyacetanilide ≥98.0%
2'-Ethoxyacetanilide
CAS Number: 581-08-8
MDL Number: MFCD00059228
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: N-Acetyl-2-ethoxyaniline, 2'-Ethoxyacetanilide
Formula:
C₁₀H₁₃NO₂ MW: 179.22 g/mol Melting Pt: 79 °C |
MDL Number:
MFCD00059228 CAS Number: 581-08-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...