4-Chlorophenylhydrazine sulfate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

C0258-25G C0258-500G
TCC0258-025GEA 48 USD
TCC0258-025G TCC0258-500G
4-Chlorophenylhydrazine sulfate ≥95.0% (by titrimetric analysis)
4-Chlorophenylhydrazine sulfate

CAS Number: 14581-21-6
MDL Number: MFCD00043829
Molecular Formula: C6H7ClN2
Molecular Weight: 383.24
Purity/Analysis Method: >95.0% (T)
Form: Crystal

Formula: C₆H₇ClN₂
MDL Number: MFCD00043829
CAS Number: 14581-21-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecf8d860a9c0b959681e263859bca574a2fd76f8&vid=ecf8d860a9c0b959681e263859bca574a2fd76f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR