1-Chloro-2-ethoxybenzene ≥99.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-ethoxybenzene, 2-Chlorophenetole

C0238-25ML C0238-10ML
TCC0238-25MLEA 218.6 USD
TCC0238-25ML TCC0238-010ML
1-Chloro-2-ethoxybenzene ≥99.0%
1-Chloro-2-ethoxybenzene

CAS Number: 614-72-2
MDL Number: MFCD00058930
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 1-Chloro-2-ethoxybenzene

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 210 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00058930
CAS Number: 614-72-2
EINECS: 210-392-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f33c94dc66d4a252d15af517263351db596b2925&vid=f33c94dc66d4a252d15af517263351db596b2925&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR