(4-Chloro-2-methylphenoxy)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C0206-25G C0206-500G
TCC0206-025GEA 52.52 USD
TCC0206-025G TCC0206-500G
(4-Chloro-2-methylphenoxy)acetic acid ≥98.0% (by titrimetric analysis)
(4-Chloro-2-methylphenoxy)acetic acid

CAS Number: 94-74-6
MDL Number: MFCD00004306
Molecular Formula: C9H9ClO3
Molecular Weight: 200.62
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 119


Product Categories: Bioscience, Herbicide Ingredients, Auxins
Other Product Names / Synonyms: MCPA

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
Melting Pt: 119 °C
MDL Number: MFCD00004306
CAS Number: 94-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2df4811adb5c5775729f958244a6eab2463dd5cf&vid=2df4811adb5c5775729f958244a6eab2463dd5cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR