N-Chloroacetyl-L-leucine ≥99.0%

Supplier: TCI America

C0101-25G C0101-1G
TCC0101-025GEA 283 USD
TCC0101-025G TCC0101-001G
N-Chloroacetyl-L-leucine ≥99.0%
N-Chloroacetyl-L-leucine

CAS Number: 688-12-0
MDL Number: MFCD00018912
Molecular Formula: C8H14ClNO3
Molecular Weight: 207.65
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 134
Specific rotation [a]20/D: -15.5 deg (C=2, EtOH)

Formula: C₈H₁₄ClNO₃
Melting Pt: 134 °C
MDL Number: MFCD00018912
CAS Number: 688-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33cba0ad3b736217ae6de5c1557111c044218d77&vid=33cba0ad3b736217ae6de5c1557111c044218d77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR