Chloranilic acid lanthanum(III) salt decahydrate ≥98.0%

Supplier: TCI America

Synonyms: Lanthanum(III) Chloranilate

C0079-5G C0079-25G
TCC0079-005GEA 57 USD
TCC0079-005G TCC0079-025G
Chloranilic acid lanthanum(III) salt decahydrate ≥98.0%
Chloranilic acid lanthanum(III) salt decahydrate

CAS Number: 32607-23-1
MDL Number: MFCD00135764
Molecular Formula: C6H2Cl2O4
Molecular Weight: 898.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Lanthanum(III) Chloranilate

Formula: C₆H₂Cl₂O₄
MW: 898.70 g/mol
MDL Number: MFCD00135764
CAS Number: 32607-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=441e5fdefa7be64a602fda32502c69119a68158e&vid=441e5fdefa7be64a602fda32502c69119a68158e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR