6-Amino-2-mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A0682-5G A0682-25G
TCA0682-005GEA 211 USD
TCA0682-005G TCA0682-025G
6-Amino-2-mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)
6-Amino-2-mercaptobenzothiazole

CAS Number: 7442-07-1
MDL Number: MFCD00059164
Molecular Formula: C7H6N2S2
Molecular Weight: 182.26
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal

Formula: C₇H₆N₂S₂
MW: 182.26 g/mol
MDL Number: MFCD00059164
CAS Number: 7442-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76e49e25466af13aacc7d16408e3ec829e7cce56&vid=76e49e25466af13aacc7d16408e3ec829e7cce56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR