Decanoic anhydride ≥98.0%

Supplier: TCI America

C0020-25G C0020-100G
TCC0020-025GEA 99 USD
TCC0020-025G TCC0020-100G
Decanoic anhydride ≥98.0%
Decanoic anhydride

CAS Number: 2082-76-0
MDL Number: MFCD00010460
Molecular Formula: C20H38O3
Molecular Weight: 326.52
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 24

Formula: C₂₀H₃₈O₃
Flash Pt: 113 °C
MDL Number: MFCD00010460
CAS Number: 2082-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e693aa8e74a6fc89ac4f0d6e2a9cd7a73ff0c3d2&vid=e693aa8e74a6fc89ac4f0d6e2a9cd7a73ff0c3d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR