(±)-Camphor-10-sulfonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: DL-Camphor-10-sulphonic acid, (±)-2-Oxobornane-10-sulphonic acid, (7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid

C0016-25G C0016-500G
TCC0016-025GEA 41.08 USD
TCC0016-025G TCC0016-500G
(±)-Camphor-10-sulfonic acid ≥98.0% (by titrimetric analysis)
(±)-Camphor-10-sulphonic acid

CAS Number: 5872-08-2
MDL Number: MFCD00074827
Molecular Formula: C10H16O4S
Molecular Weight: 232.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 202


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis, Bicyclic Monoterpenes
Other Product Names / Synonyms: (+/-)-CSA

Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00074827
CAS Number: 5872-08-2
EINECS: 227-527-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f52a279cc7edf14d4135d26bcecea3242bfab57&vid=5f52a279cc7edf14d4135d26bcecea3242bfab57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR