(±)-Camphorquinone ≥98.0%

Supplier: TCI America

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

C0014-25G C0014-5G
TCC0014-025GEA 230.43 USD
TCC0014-025G TCC0014-005G
(±)-Camphorquinone ≥98.0%
(±)-Camphorquinone

CAS Number: 10373-78-1
MDL Number: MFCD00064160
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 203


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: 2,3-Bornanedione

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00064160
CAS Number: 10373-78-1
EINECS: 233-814-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b29934255f62cd4f0e129274fcae1cd415a4dc7&vid=6b29934255f62cd4f0e129274fcae1cd415a4dc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR