DL-Camphor ≥95.0%
Supplier: TCI America
|
Warning
|
Synonyms:
DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone
C0011-25G
C0011-500G
TCC0011-025GEA
18.23
USD
TCC0011-025G
TCC0011-500G
DL-Camphor ≥95.0%
DL-Camphor
CAS Number: 76-22-2
MDL Number: MFCD00074738
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 174
Flash Point (°C): 66
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Boiling Pt: 204 °C (1013 hPa) Melting Pt: 174 °C Density: 0.992 g/cm³ (25 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074738 CAS Number: 76-22-2 EINECS: 200-945-0 UN: 2717 ADR: 4.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...