Acriflavine hydrochloride ≥90.0% (by HPLC)

Supplier: TCI America

A0677-25G A0677-100G
TCA0677-025GEA 64 USD
TCA0677-025G TCA0677-100G
Acriflavine hydrochloride ≥90.0% (by HPLC)
Acriflavine hydrochloride

CAS Number: 69235-50-3
MDL Number: MFCD00069039
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal

Formula: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
MDL Number: MFCD00069039
CAS Number: 8063-24-9

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR