2-(Bromoacetyl)-6-methoxynaphthalene ≥98.0%

Supplier: TCI America

Synonyms: 2-Bromo-6'-methoxy-2'-acetonaphthone

B3496-1G B3496-5G
TCB3496-1GEA 67 USD
TCB3496-1G TCB3496-5G
2-(Bromoacetyl)-6-methoxynaphthalene ≥98.0%
2-(Bromoacetyl)-6-methoxynaphthalene

CAS Number: 10262-65-4
MDL Number: MFCD03964189
Molecular Formula: C13H11BrO2
Molecular Weight: 279.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 112


Other Product Names / Synonyms: 2-Bromo-6'-methoxy-2'-acetonaphthone

Formula: C₁₃H₁₁BrO₂
MW: 279.13 g/mol
Melting Pt: 112 °C
MDL Number: MFCD03964189
CAS Number: 10262-65-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6256ef0aa32eac552a536b466061c44a52b0f16b&vid=6256ef0aa32eac552a536b466061c44a52b0f16b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR