2,3-Bis(4-bromophenyl)-2-butenedinitrile ≥98.0%

Supplier: TCI America

B3437-5G B3437-1G
TCB3437-5GEA 334.8 USD
TCB3437-5G TCB3437-1G
2,3-Bis(4-bromophenyl)-2-butenedinitrile ≥98.0%
2,3-Bis(4-bromophenyl)-2-butenedinitrile

CAS Number: 315203-26-0 Molecular Formula: C16H8Br2N2 Molecular Weight: 388.06 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 218

Formula: C₁₆H₈Br₂N₂
MW: 388.06 g/mol
Melting Pt: 218 °C
CAS Number: 315203-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6036ae4a20f7a43472a5f939ca0aad1f349bb38c&vid=6036ae4a20f7a43472a5f939ca0aad1f349bb38c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR