Bis(2-ethylhexyl)-3,3'-thiodipropionate ≥96.0%
Supplier: TCI America
CAS Number: 10526-15-5
MDL Number: MFCD01757653
Molecular Formula: C22H42O4S
Molecular Weight: 402.63
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 3,3'-Thiodipropionic Acid Bis(2-ethylhexyl) Ester
Formula:
C₂₂H₄₂O₄S Boiling Pt: 180 °C Density: 0.97 (20 °C) |
MDL Number:
MFCD01757653 CAS Number: 10526-15-5 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...