tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: tert-Butyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, N-Boc-Nortropinone

B3401-5G B3401-1G
TCB3401-5GEA 137 USD
TCB3401-5G TCB3401-1G
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate ≥98.0% (by HPLC, total nitrogen)
tert-Butyl-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate

CAS Number: 185099-67-6
MDL Number: MFCD00673779
Molecular Formula: C12H19NO3
Molecular Weight: 225.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 71


Product Categories: Bioscience, Tropane Alkaloids
Other Product Names / Synonyms: N-Boc-nortropinone, N-(tert-Butoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one

Formula: C₁₂H₁₉NO₃
MW: 225.29 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00673779
CAS Number: 185099-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d005baaac4157f7beb7b5509767f73997916194f&vid=d005baaac4157f7beb7b5509767f73997916194f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR