2-Bromo-α,α,α,4-tetrafluorotoluene ≥98.0%
Supplier: TCI America
Synonyms:
2-Bromo-4-fluorobenzotrifluoride, 1-Bromo-3-fluoro-6-(trifluoromethyl)benzene
B3397-25G
B3397-5G
TCB3397-25GEA
195
USD
TCB3397-25G
TCB3397-5G
2-Bromo-α,α,α,4-tetrafluorotoluene ≥98.0%
2-Bromo-α,α,α,4-tetrafluorotoluene
CAS Number: 351003-21-9
MDL Number: MFCD03094041
Molecular Formula: C7H3BrF4
Molecular Weight: 243.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 78
Specific Gravity (20/20): 1.75
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Bromo-4-fluoro-1-(trifluoromethyl)benzene, 2-Bromo-alpha,alpha,alpha,4-tetrafluorotoluene
Formula:
C₇H₃BrF₄ MW: 243 g/mol Boiling Pt: 162 °C Density: 1.75 (20 °C) Flash Pt: 78 °C |
MDL Number:
MFCD03094041 CAS Number: 351003-21-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...