Dibutyl adipate ≥99.0%

Supplier: TCI America

Synonyms: Adipic Acid Dibutyl Ester

A0655-500ML A0655-25ML
TCA0655-500MLEA 80.41 USD
TCA0655-500ML TCA0655-025ML
Dibutyl adipate ≥99.0%
Dibutyl adipate

CAS Number: 105-99-7
MDL Number: MFCD00009451
Molecular Formula: C14H26O4
Molecular Weight: 258.36
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 305
Melting point (°C): -32
Flash Point (°C): 166
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Adipic Acid Dibutyl Ester

Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
Boiling Pt: 305 °C
Melting Pt: –32 °C
Density: 0.96 (20 °C)
Flash Pt: 166 °C
MDL Number: MFCD00009451
CAS Number: 105-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=712c6f87dfdc3e316f357ee49201014a140298fd&vid=712c6f87dfdc3e316f357ee49201014a140298fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR