Bezafibrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-(4-(2-(4-Chlorobenzamido)ethyl)phenoxy)-2-methylpropanoic acid, 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid
B3346-25G
B3346-5G
TCB3346-25GEA
279
USD
TCB3346-25G
TCB3346-5G
Bezafibrate ≥98.0% (by HPLC, titration analysis)
Bezafibrate
CAS Number: 41859-67-0
MDL Number: MFCD00078970
Molecular Formula: C19H20ClNO4
Molecular Weight: 361.82
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 184
Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]isobutyric Acid
Formula:
C₁₉H₂₀ClNO₄ MW: 361.82 g/mol Melting Pt: 184 °C |
MDL Number:
MFCD00078970 CAS Number: 41859-67-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...