6-Bromochromone ≥98.0%

Supplier: TCI America

Synonyms: 6-Bromo-4H-chromen-4-one

B3335-25G B3335-5G
TCB3335-25GEA 271 USD
TCB3335-25G TCB3335-5G
6-Bromochromone ≥98.0%
6-Bromochromone

CAS Number: 51483-92-2
MDL Number: MFCD00239369
Molecular Formula: C9H5BrO2
Molecular Weight: 225.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 6-Bromo-4-oxo-4H-1-benzopyran

Formula: C₉H₅BrO₂
MW: 225.0388 g/mol
Melting Pt: 138 °C
MDL Number: MFCD00239369
CAS Number: 51483-92-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f74fad0117c89f81ae7b83dd86a4bf04796936d&vid=8f74fad0117c89f81ae7b83dd86a4bf04796936d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR