4-Benzyloxy-1,3-butanediol ≥97.0%

Supplier: TCI America

B3326-5G B3326-1G
TCB3326-5GEA 1020 USD
TCB3326-5G TCB3326-1G
4-Benzyloxy-1,3-butanediol ≥97.0%
4-Benzyloxy-1,3-butanediol

CAS Number: 71998-70-4
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 75

Formula: C₁₁H₁₆O₃
Melting Pt: 75 °C
CAS Number: 71998-70-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=253be0d148ae3af66d989f7388e415b1d9b9c248&vid=253be0d148ae3af66d989f7388e415b1d9b9c248&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR