5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Fmoc-Glu(OtBu)-OPfp, Fmoc-Glu(OtBu)-Opfp, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, N-Fmoc-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, 5-tert-Butyl 1-Pentafluorophenyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester

B3318-5G B3318-1G
TCB3318-5GEA 175 USD
TCB3318-5G TCB3318-1G
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate ≥98.0% (by HPLC, total nitrogen)
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate

CAS Number: 86061-04-3
MDL Number: MFCD00065647
Molecular Formula: C30H26F5NO6
Molecular Weight: 591.53
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 124
Specific rotation [a]20/D: -19 deg (C=1, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)

Formula: C₃₀H₂₆F₅NO₆
MW: 591.53 g/mol
Melting Pt: 124 °C
MDL Number: MFCD00065647
CAS Number: 86061-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0bbe750a2cf4eff81bb6ee42ab9766b6b0608e2&vid=b0bbe750a2cf4eff81bb6ee42ab9766b6b0608e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR