5-(4-Bromophenyl)-2-furaldehyde ≥94.0%
Supplier: TCI America
Synonyms:
5-(4-Bromophenyl)furan-2-carbaldehyde, 5-(4-Bromophenyl)furfural
B3295-5G
B3295-1G
TCB3295-5GEA
128
USD
TCB3295-5G
TCB3295-1G
5-(4-Bromophenyl)-2-furaldehyde ≥94.0%
5-(4-Bromophenyl)-2-furaldehyde
CAS Number: 20005-42-9
MDL Number: MFCD00239387
Molecular Formula: C11H7BrO2
Molecular Weight: 251.08
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Melting point (°C): 153
Other Product Names / Synonyms: 5-(4-Bromophenyl)furfural
Formula:
C₁₁H₇BrO₂ MW: 251.08 g/mol Melting Pt: 153 °C |
MDL Number:
MFCD00239387 CAS Number: 20005-42-9 |
Specification Test Results
Purity/Analysis Method | >94.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...