N-Boc-DL-phenylalaninol ≥96.0%

Supplier: TCI America

Synonyms: Boc-DL-Phenylalaninol, tert-Butyl (1-hydroxy-3-phenylpropan-2-yl)carbamate

B3269-1G B3269-5G
TCB3269-1GEA 56.5 USD
TCB3269-1G TCB3269-5G
N-Boc-DL-phenylalaninol ≥96.0%
N-Boc-DL-phenylalaninol

CAS Number: 145149-48-0
MDL Number: MFCD01457313
Molecular Formula: C14H21NO3
Molecular Weight: 251.33
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 86


Other Product Names / Synonyms: N-Boc-DL-phenylalaninol, 2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol

Formula: C₁₄H₂₁NO₃
MW: 251.3 g/mol
Melting Pt: 86 °C
MDL Number: MFCD01457313
CAS Number: 145149-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98edd944575c51038d9609bf86568a1d30fcfcd0&vid=98edd944575c51038d9609bf86568a1d30fcfcd0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR