1-(2-Bromoethoxy)-4-nitrobenzene ≥97.0%

Supplier: TCI America

B3254-1G B3254-5G
TCB3254-1GEA 130 USD
TCB3254-1G TCB3254-5G
1-(2-Bromoethoxy)-4-nitrobenzene ≥97.0%
1-(2-Bromoethoxy)-4-nitrobenzene

CAS Number: 13288-06-7
MDL Number: MFCD00031363
Molecular Formula: C8H8BrNO3
Molecular Weight: 246.06
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 66


Other Product Names / Synonyms: 1-(2-Bromoethoxy)-4-nitrobenzene, 2-Bromoethyl 4-Nitrophenyl Ether

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Boiling Pt: 170 °C
Melting Pt: 66 °C
MDL Number: MFCD00031363
CAS Number: 13288-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d19c394fa1d406d93b1cfe4d9312540330424d8&vid=8d19c394fa1d406d93b1cfe4d9312540330424d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR