4-(Bromomethyl)-3-fluorobenzonitrile ≥98.0%
Supplier: TCI America
B3204-1G
B3204-5G
TCB3204-1GEA
32
USD
TCB3204-1G
TCB3204-5G
4-(Bromomethyl)-3-fluorobenzonitrile ≥98.0%
4-(Bromomethyl)-3-fluorobenzonitrile
CAS Number: 105942-09-4
MDL Number: MFCD07368341
Molecular Formula: C8H5BrFN
Molecular Weight: 214.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-2-fluorobenzyl Bromide
Formula:
C₈H₅BrFN MW: 214.04 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD07368341 CAS Number: 105942-09-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...