5-Bromo-2-adamantanone ≥98.0%

Supplier: TCI America

Synonyms: 5-Bromoadamantan-2-one

B3154-1G B3154-5G
TCB3154-1GEA 106 USD
TCB3154-1G TCB3154-5G
5-Bromo-2-adamantanone ≥98.0%
5-Bromo-2-adamantanone

CAS Number: 20098-20-8
MDL Number: MFCD01321097
Molecular Formula: C10H13BrO
Molecular Weight: 229.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153

Formula: C₁₀H₁₃BrO
MW: 229.12 g/mol
Melting Pt: 153 °C
MDL Number: MFCD01321097
CAS Number: 20098-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=840221899aeadf9b7f7e2b111b7a9b724cbf5005&vid=840221899aeadf9b7f7e2b111b7a9b724cbf5005&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR