4-Bromo-N,N-3-trimethylaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromo-N,N-dimethyl-m-toluidine

B3133-25G B3133-5G
TCB3133-25GEA 353 USD
TCB3133-25G TCB3133-5G
4-Bromo-N,N-3-trimethylaniline ≥98.0% (by GC, titration analysis)
4-Bromo-N,N-3-trimethylaniline

CAS Number: 50638-50-1
MDL Number: MFCD01851099
Molecular Formula: C9H12BrN
Molecular Weight: 214.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 54


Other Product Names / Synonyms: 4-Bromo-N,N-dimethyl-m-toluidine

Formula: C₉H₁₂BrN
MW: 214.11 g/mol
Boiling Pt: 148 °C
Melting Pt: 54 °C
MDL Number: MFCD01851099
CAS Number: 50638-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6aa2826a3dac97773130378cafe414db22575dd&vid=c6aa2826a3dac97773130378cafe414db22575dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR