N-ɑ-Fmoc-N-ε-boc-L-lysine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B3072-5G
B3072-25G
TCB3072-5GEA
30.53
USD
TCB3072-5G
TCB3072-25G
N-ɑ-Fmoc-N-ε-boc-L-lysine ≥98.0% (by HPLC, titration analysis)
N-alpha-Fmoc-N-epsilon-boc-L-lysine
CAS Number: 71989-26-9
MDL Number: MFCD00037138
Molecular Formula: C26H32N2O6
Molecular Weight: 468.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 124
Specific rotation [a]20/D: -12 deg (C=1, DMF)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nepsilon-Boc-Nalpha-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH
Formula:
C₂₆H₃₂N₂O₆ Melting Pt: 124 °C |
MDL Number:
MFCD00037138 CAS Number: 71989-26-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...