4-Bromo-N,N-3,5-tetramethylaniline ≥98.0%

Supplier: TCI America

B3070-1G B3070-5G
TCB3070-1GEA 117 USD
TCB3070-1G TCB3070-5G
4-Bromo-N,N-3,5-tetramethylaniline ≥98.0%
4-Bromo-N,N-3,5-tetramethylaniline

CAS Number: 14275-09-3
Molecular Formula: C10H14BrN
Molecular Weight: 228.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Bromo-5-(dimethylamino)-m-xylene, 4-Bromo-N,N-dimethyl-3,5-xylidine

Formula: C₁₀H₁₄BrN
MDL Number: MFCD11036245
CAS Number: 14275-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a548f8383bfbcd6e38eb0d70a7785b8241b9984&vid=4a548f8383bfbcd6e38eb0d70a7785b8241b9984&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR