N-(tert-Butoxycarbonyl)-2,2-dimethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(3-Amino-2,2-dimethylpropyl)carbamic Acid tert-Butyl Ester, N-Boc-2,2-dimethyl-1,3-diaminopropane, tert-Butyl (3-amino-2,2-dimethylpropyl)carbamate, N-Boc-2,2-dimethyl-1,3-propanediamine
CAS Number: 292606-35-0
MDL Number: MFCD01076212
Molecular Formula: C10H22N2O2
Molecular Weight: 202.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 74
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-Boc-2,2-dimethyl-1,3-propanediamine, tert-Butyl (3-Amino-2,2-dimethylpropyl)carbamate, (3-Amino-2,2-dimethylpropyl)carbamic Acid tert-Butyl Ester, N-Boc-2,2-dimethyl-1,3-diaminopropane
Formula:
C₁₀H₂₂N₂O₂ MW: 202.30 g/mol Melting Pt: 74 °C |
MDL Number:
MFCD01076212 CAS Number: 292606-35-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...