1-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: D-Glutamic Acid 1-Benzyl Ester, H-D-Glu-OBzl

B2995-1G B2995-5G
TCB2995-1GEA 117.2 USD
TCB2995-1G TCB2995-5G
1-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-D-glutamate

CAS Number: 79338-14-0
MDL Number: MFCD00237065
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144
Specific rotation [a]20/D: -11 deg (C=1, 1mol/L HCl)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: D-Glutamic Acid 1-Benzyl Ester, H-D-Glu-OBzl

Formula: C₁₂H₁₅NO₄
Melting Pt: 144 °C
MDL Number: MFCD00237065
CAS Number: 79338-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d12f7eaf3ee512b4224374ca5d4b3a8195d7a7&vid=7dc8e6f918628193468e9183cbc9451138f597b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR