1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: L-Aspartic Acid 1-Benzyl Ester, H-Asp-OBzl

B2994-5G B2994-1G
TCB2994-5GEA 39 USD
TCB2994-5G TCB2994-1G
1-Benzyl-L-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-L-aspartate

CAS Number: 7362-93-8
MDL Number: MFCD00063186
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 166
Specific rotation [a]20/D: -14 deg (C=1, 0.1mol/L HCl)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartic Acid 1-Benzyl Ester, H-Asp-OBzl

Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00063186
CAS Number: 7362-93-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d19e6fa0b7a9845905b274f78043909816d7da59&vid=d19e6fa0b7a9845905b274f78043909816d7da59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR