2-(3-Benzoylphenyl)propionitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-(3-Benzoylphenyl)propanenitrile, Ketoprofen Related Compound F, 3-Benzoyl-alpha-methylphenylacetonitrile, 3-(1-Cyanoethyl)benzophenone

B2939-25G B2939-5G
TCB2939-25GEA 199 USD
TCB2939-25G TCB2939-5G
2-(3-Benzoylphenyl)propionitrile ≥98.0%
2-(3-Benzoylphenyl)propionitrile

CAS Number: 42872-30-0
MDL Number: MFCD00044435
Molecular Formula: C16H13NO
Molecular Weight: 235.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 113


Other Product Names / Synonyms: 3-(1-Cyanoethyl)benzophenone, 3-Benzoyl-alpha-methylphenylacetonitrile

Formula: C₁₆H₁₃NO
MW: 235.29 g/mol
Melting Pt: 57 °C
Flash Pt: 113 °C
MDL Number: MFCD00044435
CAS Number: 42872-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f649826f4967a8b01a27b920db8ba4ed667ecd0e&vid=f649826f4967a8b01a27b920db8ba4ed667ecd0e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR