(S)-4-Benzyloxy-1,2-butanediol ≥95.0%
Supplier: TCI America
B2900-1G
B2900-200MG
TCB2900-1GEA
372.4
USD
TCB2900-1G
TCB2900-200MG
(S)-4-Benzyloxy-1,2-butanediol ≥95.0%
(S)-4-Benzyloxy-1,2-butanediol
CAS Number: 69985-32-6
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: -23 deg (C=5, EtOH)
Formula:
C₁₁H₁₆O₃ Boiling Pt: 170 °C Density: 1.12 (20 °C) |
MDL Number:
MFCD10000950 CAS Number: 69985-32-6 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...