Benzyloxyacetic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
B2876-1G
B2876-5G
TCB2876-1GEA
22
USD
TCB2876-1G
TCB2876-5G
Benzyloxyacetic acid ≥97.0% (by GC, titration analysis)
Benzyloxyacetic acid
CAS Number: 30379-55-6
MDL Number: MFCD00274211
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 113
Specific Gravity (20/20): 1.17
Formula:
C₉H₁₀O₃ Boiling Pt: 155 °C Density: 1.17 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00274211 CAS Number: 30379-55-6 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...