2-(4-Bromophenyl)benzimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

B2812-5G B2812-1G
TCB2812-5GEA 317 USD
TCB2812-5G TCB2812-1G
2-(4-Bromophenyl)benzimidazole ≥98.0% (by HPLC, titration analysis)
2-(4-Bromophenyl)benzimidazole

CAS Number: 2622-74-4
Molecular Formula: C13H9BrN2
Molecular Weight: 273.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 299

Formula: C₁₃H₉BrN₂
Melting Pt: 299 °C
MDL Number: MFCD00224360
CAS Number: 2622-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c94fdde4e2deb33334f51ab298156a3dfd3168ef&vid=c94fdde4e2deb33334f51ab298156a3dfd3168ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR