(4-Bromophenoxy)acetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Bromophenoxyacetic acid
B2746-25G
B2746-5G
TCB2746-25GEA
68
USD
TCB2746-25G
TCB2746-5G
(4-Bromophenoxy)acetic acid ≥98.0% (by titrimetric analysis)
(4-Bromophenoxy)acetic acid
CAS Number: 1878-91-7
Molecular Formula: C8H7BrO3
Molecular Weight: 231.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 159
Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 4-BPA
Formula:
(C₆H₄Br)OCH₂COOH MW: 231.05 g/mol Melting Pt: 159 °C |
MDL Number:
MFCD00093070 CAS Number: 1878-91-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...