Bis(4-iodophenyl)amine ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Diiododiphenylamine

B2685-1G B2685-5G
TCB2685-1GEA 81 USD
TCB2685-1G TCB2685-5G
Bis(4-iodophenyl)amine ≥98.0%
Bis(4-iodophenyl)amine

CAS Number: 20255-70-3
Molecular Formula: C12H9I2N
Molecular Weight: 421.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4,4'-Diiododiphenylamine

Formula: C₁₂H₉I₂N
MW: 421.02 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00454093
CAS Number: 20255-70-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=260c042b1988099ca7e1ed255890df718c248dde&vid=260c042b1988099ca7e1ed255890df718c248dde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR