2-(4-Bromophenyl)benzothiazole ≥98.0%
Supplier: TCI America
Synonyms:
1-(2-Benzothiazolyl)-4-bromobenzene
B2679-5G
B2679-25G
TCB2679-5GEA
85.8
USD
TCB2679-5G
TCB2679-25G
2-(4-Bromophenyl)benzothiazole ≥98.0%
2-(4-Bromophenyl)benzothiazole
CAS Number: 19654-19-4
Molecular Formula: C13H8BrNS
Molecular Weight: 290.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 134
Other Product Names / Synonyms: 1-(2-Benzothiazolyl)-4-bromobenzene
Formula:
C₁₃H₈BrNS MW: 290.18 g/mol Melting Pt: 134 °C |
MDL Number:
MFCD00579199 CAS Number: 19654-19-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...