3-Bromo-2,4-dimethyl-5-phenylthiophene ≥98.0%
Supplier: TCI America
B2677-1G
B2677-5G
TCB2677-1GEA
99
USD
TCB2677-1G
TCB2677-5G
3-Bromo-2,4-dimethyl-5-phenylthiophene ≥98.0%
3-Bromo-2,4-dimethyl-5-phenylthiophene
CAS Number: 362513-28-8
Molecular Formula: C12H11BrS
Molecular Weight: 267.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 32
Storage Temperature: 0-10°C
Formula:
C₁₂H₁₁BrS |
MDL Number:
MFCD08276302 CAS Number: 362513-28-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...