Acid orange 5

Supplier: TCI America

Synonyms: OrangeIV, Aniline yellow, 4-(4-Anilinophenylazo)benzenesulphonic acid sodium salt, CI 13080, Diphenylamine orange, Indicator, S. No.: 179, Orange IV, Tropaeolin OO

A0576-25G A0576-100G
TCA0576-025GEA 57 USD
TCA0576-025G TCA0576-100G
Acid orange 5
Acid orange 5

CAS Number: 554-73-4 MDL Number: MFCD00038130 Molecular Formula: C18H15N3O3S Molecular Weight: 375.38 Form: Crystal Color: Yellow


Form: Crystal
Color: Yellow

Formula: C₁₈H₁₄N₃NaO₃S
MW: 375.38 g/mol
MDL Number: MFCD00038130
CAS Number: 554-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea8db94153a3cfd89d650ad7285f74a51d49e233&vid=ea8db94153a3cfd89d650ad7285f74a51d49e233&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR