Bis(4-tert-butylphenyl)iodonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Bis(4-(tert-butyl)phenyl)iodonium hexafluorophosphate(V)

B2380-5G B2380-1G
TCB2380-5GEA 362 USD
TCB2380-5G TCB2380-1G
Bis(4-tert-butylphenyl)iodonium hexafluorophosphate ≥98.0% (by HPLC, titration analysis)
Bis(4-tert-butylphenyl)iodonium hexafluorophosphate

CAS Number: 61358-25-6
MDL Number: MFCD06797070
Molecular Formula: C20H26F6IP
Molecular Weight: 538.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 175

Formula: C₂₀H₂₆F₆IP
MW: 538.30 g/mol
Melting Pt: 175 °C
MDL Number: MFCD06797070
CAS Number: 61358-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32e8aa5bb6bbf63f62237643c9c33ac69c6bdbe&vid=e32e8aa5bb6bbf63f62237643c9c33ac69c6bdbe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR