(1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0%
Supplier: TCI America
Synonyms:
(1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine
B2316-100MG
B2316-500MG
TCB2316-100MGEA
197
USD
TCB2316-100MG
TCB2316-500MG
(1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine ≥97.0%
(1R,2R)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine
CAS Number: 425615-42-5
MDL Number: MFCD06797063
Molecular Formula: C20H28N2
Molecular Weight: 296.46
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: 203 deg (C=1, CH2Cl2)
Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Chiral Building Blocks
Other Product Names / Synonyms: (1R,2R)-1,2-Diamino-1,2-dimesitylethane, (1R,2R)-1,2-Dimesitylethylenediamine
Formula:
C₂₀H₂₈N₂ MW: 296.46 g/mol Melting Pt: 86 °C |
MDL Number:
MFCD06797063 CAS Number: 425615-42-5 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...