N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Boc-2-nitrobenzenesulphonamide

B2303-1G B2303-25G B2303-5G
TCB2303-1GEA 58 USD
TCB2303-1G TCB2303-25G TCB2303-5G
N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-2-nitrobenzenesulfonamide

CAS Number: 198572-71-3
MDL Number: MFCD06796217
Molecular Formula: C11H14N2O6S
Molecular Weight: 302.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Amination, Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-Boc-2-nitrobenzenesulfonamide

Formula: C₁₁H₁₄N₂O₆S
MW: 302.30 g/mol
MDL Number: MFCD06796217
CAS Number: 198572-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc0b983c1bdecfdec4ab8f1d04e7d185bbe47a7&vid=efc0b983c1bdecfdec4ab8f1d04e7d185bbe47a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR