Atropine sulfate Monohydrate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

A0550-5G A0550-25G
TCA0550-005GEA 33.45 USD
TCA0550-005G TCA0550-025G
Atropine sulfate Monohydrate ≥99.0% (by titrimetric analysis)
Atropine sulfate monohydrate

CAS Number: 5908-99-6
MDL Number: MFCD00077268
Molecular Formula: C17H23NO3
Molecular Weight: 676.82
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 194

Formula: C₁₇H₂₃NO₃
MW: 676.82 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00077268
CAS Number: 5908-99-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbee1dadecf69aa2498c2ff287f6b299d412974a&vid=dbee1dadecf69aa2498c2ff287f6b299d412974a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR